Assay Details
| Gene Symbol: Car3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019292.4 | NP_062165.2 | 3 - 4 | 356 | 80 | |
| XM_006232116.1 | XP_006232178.1 | 4 - 5 | 681 | 80 | ||
| GenBank mRNA | AB030829.1 | - | 3 - 4 | 356 | 80 | |
| AB030830.1 | - | 3 - 4 | 356 | 80 | ||
| AF037072.1 | - | 3 - 4 | 379 | 80 | ||
| BC061980.1 | - | 3 - 4 | 391 | 5597975 | 80 | |
| FQ210528.1 | - | 3 - 4 | 404 | 80 | ||
| FQ211459.1 | - | 3 - 4 | 403 | 80 | ||
| FQ214846.1 | - | 3 - 4 | 401 | 80 | ||
| FQ214929.1 | - | 3 - 4 | 406 | 80 | ||
| FQ216182.1 | - | 3 - 4 | 402 | 80 | ||
| FQ223846.1 | - | 3 - 4 | 402 | 80 | ||
| M22413.1 | - | 3 - 4 | 511 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car3
Genomic Map