Assay Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133399.2 | NP_596890.2 | 10 - 11 | 1747 | 124 | |
| GenBank mRNA | FM096453.1 | - | 2 - 3 | 211 | 124 | |
| Gene Symbol: LOC685590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 1873 | 124 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pik3ca
Genomic Map