Assay Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133399.2 | NP_596890.2 | 19 - 20 | 2934 | 118 | |
| GenBank mRNA | AF395897.1 | - | 7 - 8 | 822 | 118 | |
| Gene Symbol: LOC685590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773148.1 | XP_008771370.1 | 3 | 3054 | 118 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
protein phosphorylation
positive regulation of peptidyl-serine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
protein kinase B signaling
negative regulation of neuron apoptotic process
hypomethylation of CpG island
positive regulation of protein kinase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
negative regulation of fibroblast apoptotic process
regulation of genetic imprinting
negative regulation of anoikis
ATP binding
1-phosphatidylinositol-3-kinase activity
protein kinase activator activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
glucose metabolic process
protein phosphorylation
positive regulation of peptidyl-serine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
protein kinase B signaling
negative regulation of neuron apoptotic process
hypomethylation of CpG island
positive regulation of protein kinase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
negative regulation of fibroblast apoptotic process
regulation of genetic imprinting
negative regulation of anoikis
ATP binding
1-phosphatidylinositol-3-kinase activity
protein kinase activator activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P066438 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P073033 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P562984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P111214 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P101331 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P130410 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066438 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P562984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073033 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P101331 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130410 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P111214 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073033 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P562984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130410 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P101331 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P111214 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066438 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P130410 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P073033 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P562984 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P066438 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P111214 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P101331 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P156468 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10814747 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814750 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814749 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814748 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814751 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10814726 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10937594 | - | - |
| Rat Genome 230 2.0 Array | 1374232_at | - | - |
| Rat Genome 230 2.0 Array | 1379041_at | - | - |
| Rat Genome 230 2.0 Array | 1374232_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393499_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389143_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396411_at | - | - |
| Rat Genome 230 2.0 Array | 1379041_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382366_at | - | - |
| Rat Genome 230 2.0 Array | 1382366_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389143_at | - | - |
| Rat Genome 230 2.0 Array | 1396411_at | - | - |
| Rat Genome 230 2.0 Array | 1393499_at | - | - |
| Rat Genome 230 2.0 Array | 1396411_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396411_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379041_at | - | - |
| Affymetrix HT Rat Focus Array | 1379041_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382366_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374232_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382366_at | - | - |
| Affymetrix HT Rat Focus Array | 1393499_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393499_at | - | - |
| Affymetrix HT Rat Focus Array | 1374232_at | - | - |
| Affymetrix HT Rat Focus Array | 1396411_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1396411_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200005787.rn.1 | TC0200000953.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005784.rn.1 | TC0200000953.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005789.rn.1 | TC0200000953.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005788.rn.1 | TC0200000953.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374232_PM_at | - | - |
| Rat Expression Set 230 | 1389143_at | - | - |
| Rat Expression Set 230 | 1374232_at | - | - |
| Rat Expression Set 230 | 1389143_PM_at | - | - |
| Rat Expression Set 230 | 1396411_PM_at | - | - |
| Rat Expression Set 230 | 1382366_PM_at | - | - |
| Rat Expression Set 230 | 1379041_at | - | - |
| Rat Expression Set 230 | 1379041_PM_at | - | - |
| Rat Expression Set 230 | 1393499_PM_at | - | - |
| Rat Expression Set 230 | 1396411_at | - | - |
| Rat Expression Set 230 | 1393499_at | - | - |
| Rat Expression Set 230 | 1396411_at | - | - |
| Rat Expression Set 230 | 1396411_PM_at | - | - |
| Rat Expression Set 230 | 1382366_at | - | - |
| Rat Genome U34 Set | rc_AI030797_at | - | - |
| Rat Genome U34 Set | rc_AA997895_at | - | - |
| Rat Genome U34 Set | rc_AA964375_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22066766 | - | - |

Pik3ca
Genomic Map