Assay Details
| Gene Symbol: Sept6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173429.1 | NP_001166900.1 | 2 - 3 | 199 | 58 | |
| XM_006257463.2 | XP_006257525.1 | 2 - 3 | 522 | 58 | ||
| XM_008773441.1 | XP_008771663.1 | 2 - 3 | 522 | 58 | ||
| XM_008773442.1 | XP_008771664.1 | 2 - 3 | 522 | 58 | ||
| XM_008773443.1 | XP_008771665.1 | 2 - 3 | 522 | 58 | ||
| XM_008773444.1 | XP_008771666.1 | 2 - 3 | 229 | 58 | ||
| GenBank mRNA | BC169051.1 | - | 2 - 3 | 199 | 7381285 | 58 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008773444.1 transcripts.
Set Membership: |
|

Sept6
Genomic Map