Assay Details
| Gene Symbol: Bicd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108653.1 | NP_001102123.1 | 6 - 7 | 2675 | 90 | |
| XM_006237703.2 | XP_006237765.1 | 6 - 7 | 2655 | 90 | ||
| XM_006237705.2 | XP_006237767.1 | 6 - 7 | 2655 | 90 | ||
| XM_006237706.2 | XP_006237768.1 | 6 - 7 | 2655 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bicd1
Genomic Map