Assay Details
| Gene Symbol: LOC103690024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763277.1 | XP_008761499.1 | 580 | 96 | ||
| Gene Symbol: Pex5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170584.1 | NP_001164055.1 | 5 - 6 | 648 | 96 | |
| XM_006237384.2 | XP_006237446.1 | 5 - 6 | 649 | 96 | ||
| XM_006237385.2 | XP_006237447.1 | 5 - 6 | 579 | 96 | ||
| GenBank mRNA | CK477113.1 | - | 5 - 6 | 517 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC103690024
Genomic Map