Assay Details
| Gene Symbol: LOC103694865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773813.1 | XP_008772035.1 | 413 | 69 | ||
| XM_008773814.1 | XP_008772036.1 | 585 | 69 | |||
| Gene Symbol: Taf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105824.1 | NP_001099294.1 | 5 - 6 | 406 | 69 | |
| GenBank mRNA | BC166769.1 | - | 5 - 6 | 358 | 7933997 | 69 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694865
Genomic Map