Assay Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013043.1 | NP_001013061.1 | 1 - 2 | 62 | 87 | |
| XM_006247885.2 | XP_006247947.1 | 1 - 2 | 89 | 87 | ||
| GenBank mRNA | BC078937.1 | - | 1 - 2 | 62 | 7108345 | 87 |
| FQ225276.1 | - | 1 - 2 | 55 | 87 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC078937.1,NM_001013043.1,XM_006247885.2 transcripts.
Set Membership: |
|

Sectm1a
Genomic Map