Assay Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182675.1 | NP_872616.1 | 1 - 2 | 140 | 80 | |
| XM_006245981.2 | XP_006246043.2 | 2 - 3 | 237 | 80 | ||
| GenBank mRNA | AY190240.1 | - | 1 - 2 | 140 | 80 | |
| BC128720.1 | - | 1 - 2 | 181 | 8374477 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab40c
Genomic Map