Assay Details
| Gene Symbol: Srsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109552.2 | NP_001103022.1 | 2 - 3 | 500 | 69 | |
| XM_008768114.1 | XP_008766336.1 | 1 - 2 | 595 | 69 | ||
| GenBank mRNA | FM051934.1 | - | 2 - 3 | 479 | 69 | |
| FM055064.1 | - | 2 - 3 | 453 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srsf1
Genomic Map