Assay Details
| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005876.1 | NP_001005876.1 | 1 - 2 | 161 | 133 | |
| GenBank mRNA | BC079311.1 | - | 1 - 2 | 161 | 7134794 | 133 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079311.1, NM_001005876.1 transcripts.
Set Membership: |
|

Hn1
Genomic Map