Assay Details
| Gene Symbol: Zfp213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105764.1 | NP_001099234.1 | 2 - 3 | 791 | 96 | |
| XM_006245862.2 | XP_006245924.1 | 2 - 3 | 808 | 96 | ||
| XM_006245863.2 | XP_006245925.1 | 2 - 3 | 1344 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp213
Genomic Map