Assay Details
| Gene Symbol: Hexim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025136.1 | NP_001020307.1 | 1 | 1933 | 106 | |
| GenBank mRNA | BC087133.1 | - | 1 | 1933 | 7122255 | 106 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hexim1
Genomic Map