Assay Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191655.1 | NP_001178584.1 | 2 - 3 | 203 | 101 | |
| XM_006247715.2 | XP_006247777.1 | 2 - 3 | 408 | 101 | ||
| XM_006247716.2 | XP_006247778.1 | 2 - 3 | 257 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191655.1, XM_006247715.2, XM_006247716.2 transcripts.
Set Membership: |
|

Tmem104
Genomic Map