Assay Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031591.1 | NP_113779.1 | 2078 | 112 | ||
| GenBank mRNA | BC104711.1 | - | 2078 | 7368518 | 112 | |
More Information
Set Membership: |
|

Pecam1
Genomic Map