Assay Details
| Gene Symbol: Zfp652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080207.1 | NP_001073676.1 | 2 - 3 | 1192 | 105 | |
| GenBank mRNA | BC129097.1 | - | 2 - 3 | 1192 | 8365300 | 105 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp652
Genomic Map