Assay Details
| Gene Symbol: Tnfsf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009623.1 | NP_001009623.1 | 2 - 3 | 578 | 101 | |
| XM_006246625.2 | XP_006246687.1 | 3 - 4 | 1160 | 104 | ||
| XM_008767796.1 | XP_008766018.1 | 2 - 3 | 1316 | 104 | ||
| XM_008767797.1 | XP_008766019.1 | 2 - 3 | 1316 | 101 | ||
| XM_008767800.1 | XP_008766022.1 | 2 - 3 | 1316 | 104 | ||
| XM_008767801.1 | XP_008766023.1 | 2 - 3 | 1316 | 101 | ||
| GenBank mRNA | BC087135.1 | - | 2 - 3 | 578 | 7123934 | 101 |
| FQ215356.1 | - | 2 - 3 | 348 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf13
Genomic Map