Assay Details
| Gene Symbol: Dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106050.1 | NP_001099520.1 | 1 - 2 | 259 | 65 | |
| GenBank mRNA | FQ229062.1 | - | 1 - 2 | 133 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P370385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5603 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P370385 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P370385 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P370385 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P370385 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17589 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103233 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10785376 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785373 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785374 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785375 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10785372 | - | - |
| Rat Genome 230 2.0 Array | 1373064_at | - | - |
| Rat Genome 230 2.0 Array | 1373064_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373064_at | - | - |
| Affymetrix HT Rat Focus Array | 1373064_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500014551.rn.1 | TC1500002116.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500014550.rn.1 | TC1500002116.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500014548.rn.1 | TC1500002116.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500014549.rn.1 | TC1500002116.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500014552.rn.1 | TC1500002116.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373064_PM_at | - | - |
| Rat Expression Set 230 | 1373064_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785372 | - | - |
| Rat Genome U34 Set | rc_AI179239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21909005 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360724 | - | - |

Dnajc15
Genomic Map