Assay Details
| Gene Symbol: Ost4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134690.1 | NP_001128162.1 | 1 - 2 | 313 | 68 | |
| NM_001134691.1 | NP_001128163.1 | 2 - 3 | 213 | 68 | ||
| GenBank mRNA | BC157814.1 | - | 1 - 2 | 291 | 8362186 | 68 |
| BI277067.1 | - | 1 - 2 | 68 | |||
| BI298452.1 | - | 1 - 2 | 68 | |||
| BQ194673.1 | - | 1 - 2 | 68 | |||
| CB706964.1 | - | 2 - 3 | 174 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ost4
Genomic Map