Assay Details
| Gene Symbol: Ost4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134691.1 | NP_001128163.1 | 1 - 2 | 63 | 90 | |
| GenBank mRNA | BI298452.1 | - | 2 - 3 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001134691.1 transcripts.
This assay is in the 5'UTR region of the NM_001134691.1 transcripts.
Set Membership: |
|

Ost4
Genomic Map