Assay Details
| Gene Symbol: RGD1305089 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106730.2 | NP_001100200.1 | 3 - 4 | 1679 | 63 | |
| GenBank mRNA | BC168878.1 | - | 3 - 4 | 1679 | 7382020 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1305089
Genomic Map