Assay Details
| Gene Symbol: NEWGENE_1564769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254170.2 | XP_006254232.1 | 5 - 6 | 761 | 132 | |
| XM_006254171.2 | XP_006254233.1 | 5 - 6 | 776 | 132 | ||
| XM_008771835.1 | XP_008770057.1 | 4 - 5 | 1044 | 132 | ||
| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108418.1 | NP_001101888.1 | 610 | 132 | ||
| XM_006254175.2 | XP_006254237.1 | 761 | 132 | |||
| XM_006254176.2 | XP_006254238.1 | 776 | 132 | |||
| XM_008771752.1 | XP_008769974.1 | 1044 | 132 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_1564769
Genomic Map