Assay Details
| Gene Symbol: LOC100910173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750962.3 | XP_003751010.2 | 1 - 2 | 376 | 110 | |
| Gene Symbol: Slc25a19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007674.1 | NP_001007675.1 | 2 - 3 | 210 | 110 | |
| GenBank mRNA | BC079002.1 | - | 2 - 3 | 210 | 7112815 | 110 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910173
Genomic Map