Assay Details
| Gene Symbol: Cdc42se2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767711.1 | XP_008765933.1 | 1 - 2 | 1187 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008767711.1 transcripts.
Set Membership: |
|

Cdc42se2
Genomic Map