Assay Details
| Gene Symbol: Cdc42se2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126089.2 | NP_001119561.1 | 5 - 6 | 759 | 184 | |
| XM_006246334.1 | XP_006246396.1 | 5 - 6 | 788 | 184 | ||
| XM_008767711.1 | XP_008765933.1 | 4 - 5 | 1717 | 184 | ||
| GenBank mRNA | BC129107.1 | - | 5 - 6 | 759 | 184 | |
| BC169039.1 | - | 4 - 5 | 623 | 9027524 | 184 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P098846 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P959960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176256 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098846 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P959960 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P959960 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098846 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P959960 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P098846 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68697 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10706248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742677 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742676 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742675 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10742674 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10706246 | - | - |
| Rat Genome 230 2.0 Array | 1394639_at | - | - |
| Rat Genome 230 2.0 Array | 1388448_at | - | - |
| Rat Genome 230 2.0 Array | 1388448_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392542_at | - | - |
| Rat Genome 230 2.0 Array | 1394639_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392542_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392542_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388448_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388448_at | - | - |
| Affymetrix HT Rat Focus Array | 1394639_at | - | - |
| Affymetrix HT Rat Focus Array | 1394639_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392542_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000024623.rn.1 | TC1000002813.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000024624.rn.1 | TC1000002813.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000024622.rn.1 | TC1000002813.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000024621.rn.1 | TC1000002813.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388448_PM_at | - | - |
| Rat Expression Set 230 | 1388448_at | - | - |
| Rat Expression Set 230 | 1394639_at | - | - |
| Rat Expression Set 230 | 1392542_at | - | - |
| Rat Expression Set 230 | 1394639_PM_at | - | - |
| Rat Expression Set 230 | 1392542_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10706246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742674 | - | - |
| Rat Genome U34 Set | rc_AA942942_s_at | - | - |
| Rat Genome U34 Set | rc_AI235065_i_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22156579 | - | - |

Cdc42se2
Genomic Map