Assay Details
| Gene Symbol: Cdc42se2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126089.2 | NP_001119561.1 | 2 - 3 | 233 | 67 | |
| XM_006246334.1 | XP_006246396.1 | 2 - 3 | 262 | 67 | ||
| GenBank mRNA | BC129107.1 | - | 2 - 3 | 233 | 67 | |
| BC169039.1 | - | 1 - 2 | 100 | 9027524 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC169039.1, NM_001126089.2, XM_006246334.1 transcripts.
Set Membership: |
|

Cdc42se2
Genomic Map