Assay Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107075.1 | NP_001100545.1 | 1 - 2 | 119 | 103 | |
| XM_006247890.2 | XP_006247952.1 | 1 - 2 | 516 | 103 | ||
| XM_006247891.2 | XP_006247953.1 | 1 - 2 | 516 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107075.1 transcripts.
Set Membership: |
|

Foxk2
Genomic Map