Assay Details
| Gene Symbol: Adra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016991.2 | NP_058687.2 | 1 - 2 | 1337 | 125 | |
| GenBank mRNA | BC070920.1 | - | 1 - 2 | 1337 | 7105608 | 125 |
| M60655.1 | - | 1 - 2 | 962 | 125 | ||
| X51585.1 | - | 1 - 2 | 1188 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel remodeling
response to amphetamine
vasoconstriction of artery involved in baroreceptor response to lowering of systemic arterial blood pressure
positive regulation of heart rate by epinephrine-norepinephrine
positive regulation of the force of heart contraction by epinephrine-norepinephrine
positive regulation of cytosolic calcium ion concentration
adult heart development
locomotory behavior
visual learning
response to hormone
cell growth
regulation of vasoconstriction
negative regulation of Rho protein signal transduction
organ growth
response to drug
glucose homeostasis
response to morphine
positive regulation of MAPK cascade
response to estrogen
negative regulation of glycogen catabolic process
positive regulation of glycogen catabolic process
behavioral response to cocaine
response to steroid hormone
regulation of cardiac muscle contraction
adrenergic receptor signaling pathway
alpha1-adrenergic receptor activity
protein heterodimerization activity
blood vessel remodeling
response to amphetamine
vasoconstriction of artery involved in baroreceptor response to lowering of systemic arterial blood pressure
positive regulation of heart rate by epinephrine-norepinephrine
positive regulation of the force of heart contraction by epinephrine-norepinephrine
positive regulation of cytosolic calcium ion concentration
adult heart development
locomotory behavior
visual learning
response to hormone
cell growth
regulation of vasoconstriction
negative regulation of Rho protein signal transduction
organ growth
response to drug
glucose homeostasis
response to morphine
positive regulation of MAPK cascade
response to estrogen
negative regulation of glycogen catabolic process
positive regulation of glycogen catabolic process
behavioral response to cocaine
response to steroid hormone
regulation of cardiac muscle contraction
adrenergic receptor signaling pathway
alpha1-adrenergic receptor activity
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P138011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129727 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P138011 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P138011 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P138011 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P586943 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P586939 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P451026 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700482654 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36111 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10254 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2043 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33748 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35527 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700482654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10742215 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742217 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742214 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742218 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10742213 | - | - |
| Rat Genome 230 2.0 Array | 1368574_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368574_at | - | - |
| Affymetrix HT Rat Focus Array | 1368574_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368574_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000023372.rn.1 | TC1000002621.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000023374.rn.1 | TC1000002621.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000023373.rn.1 | TC1000002621.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000023375.rn.1 | TC1000002621.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368574_at | - | - |
| Rat Expression Set 230 | 1368574_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10742213 | - | - |
| Rat Genome U34 Set | M60655_at | - | - |
| Rat Genome U34 Set | M60655_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21518266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352223 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65953 | - | - |

Adra1b
Genomic Map