Assay Details
| Gene Symbol: Sept9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113497.1 | NP_001106969.1 | 826 | 140 | ||
| NM_031837.2 | NP_114025.2 | 1714 | 140 | |||
| NM_176856.2 | NP_789826.2 | 1833 | 140 | |||
| GenBank mRNA | AF170253.1 | - | 1831 | 137 | ||
| AF173899.1 | - | 1710 | 137 | |||
| AF180525.1 | - | 993 | 140 | |||
| AF180526.1 | - | 814 | 140 | |||
| BC166602.1 | - | 1379 | 140 | |||
| Gene Symbol: LOC103693480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768440.1 | XP_008766662.1 | 1 - 2 | 331 | 140 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768440.1 transcripts.
Set Membership: |
|

Sept9
Genomic Map