Assay Details
| Gene Symbol: LOC24906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246376.2 | XP_006246438.1 | 5 - 6 | 632 | 68 | |
| XM_006246377.2 | XP_006246439.1 | 5 - 6 | 662 | 68 | ||
More Information
Set Membership: |
|

LOC24906
Genomic Map