Assay Details
| Gene Symbol: Sppl2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105847.1 | NP_001099317.1 | 1 | 2116 | 67 | |
| XM_006247504.2 | XP_006247566.1 | 1 | 2112 | 67 | ||
| GenBank mRNA | BC160911.1 | - | 1 | 2085 | 7456885 | 67 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sppl2c
Genomic Map