Assay Details
| Gene Symbol: Rad51ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239899.2 | XP_006239961.1 | 2 - 3 | 3153 | 156 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad51ap2| Gene Symbol: Rad51ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239899.2 | XP_006239961.1 | 2 - 3 | 3153 | 156 | |
Set Membership: |
|