Assay Details
| Gene Symbol: Ap4s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130999.1 | NP_001124471.1 | 2 - 3 | 329 | 69 | |
| GenBank mRNA | BC158697.1 | - | 329 | 9026544 | 69 | |
| FQ217797.1 | - | 2 - 3 | 359 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap4s1
Genomic Map