Assay Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024359.1 | NP_077335.1 | 6 - 7 | 797 | 90 | |
| XM_006240196.2 | XP_006240258.1 | 6 - 7 | 1110 | 90 | ||
| XM_006240197.2 | XP_006240259.1 | 6 - 7 | 1107 | 90 | ||
| XM_006240198.2 | XP_006240260.1 | 6 - 7 | 1106 | 90 | ||
| XM_006240199.2 | XP_006240261.1 | 6 - 7 | 809 | 90 | ||
| XM_008764695.1 | XP_008762917.1 | 6 - 7 | 894 | 90 | ||
| GenBank mRNA | AF057308.1 | - | 6 - 7 | 797 | 90 | |
| Y09507.1 | - | 6 - 7 | 894 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hif1a
Genomic Map