Assay Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239834.2 | XP_006239896.1 | 6 - 7 | 685 | 113 | |
| XM_006239836.2 | XP_006239898.2 | 6 - 7 | 679 | 113 | ||
More Information
Set Membership: |
|

Zfp512
Genomic Map