Assay Details
| Gene Symbol: Cmpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108017.1 | NP_001101487.1 | 1 - 2 | 669 | 86 | |
| XM_006239938.2 | XP_006240000.1 | 2 - 3 | 1356 | 86 | ||
| XM_006239939.2 | XP_006240001.1 | 2 - 3 | 1356 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cmpk2
Genomic Map