Assay Details
| Gene Symbol: LOC100911440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749008.3 | XP_003749056.1 | 1012 | 128 | ||
| XM_008760026.1 | XP_008758248.1 | 979 | 128 | |||
| XM_008760027.1 | XP_008758249.1 | 985 | 128 | |||
| XM_008760028.1 | XP_008758250.1 | 937 | 128 | |||
| XM_008760029.1 | XP_008758251.1 | 1540 | 128 | |||
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230557.2 | XP_006230619.1 | 9 - 10 | 1012 | 128 | |
| XM_008759993.1 | XP_008758215.1 | 8 - 9 | 979 | 128 | ||
| XM_008759994.1 | XP_008758216.1 | 8 - 9 | 985 | 128 | ||
| XM_008759995.1 | XP_008758217.1 | 8 - 9 | 937 | 128 | ||
| XM_008759996.1 | XP_008758218.1 | 8 - 9 | 1540 | 128 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911440
Genomic Map