Assay Details
| Gene Symbol: LOC100911440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760028.1 | XP_008758250.1 | 57 | 88 | ||
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759995.1 | XP_008758217.1 | 1 - 2 | 57 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008759995.1, XM_008760028.1 transcripts.
Set Membership: |
|

LOC100911440
Genomic Map