Assay Details
| Gene Symbol: LOC100911440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749008.3 | XP_003749056.1 | 134 | 73 | ||
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230557.2 | XP_006230619.1 | 2 - 3 | 134 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003749008.3, XM_006230557.2 transcripts.
Set Membership: |
|

LOC100911440
Genomic Map