Assay Details
| Gene Symbol: Slc25a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014027.1 | NP_001014049.1 | 1 - 2 | 82 | 67 | |
| XM_008759993.1 | XP_008758215.1 | 1 - 2 | 106 | 67 | ||
| GenBank mRNA | BC090010.1 | - | 1 - 2 | 82 | 7324394 | 67 |
| Gene Symbol: LOC100911440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760026.1 | XP_008758248.1 | 106 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC090010.1,NM_001014027.1,XM_008759993.1,XM_008760026.1 transcripts.
Set Membership: |
|

Slc25a22
Genomic Map