Assay Details
| Gene Symbol: Aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172327.2 | NP_758830.2 | 2 - 3 | 278 | 62 | |
| XM_006230699.2 | XP_006230761.1 | 3 - 4 | 476 | 62 | ||
| XM_006230700.2 | XP_006230762.1 | 3 - 4 | 445 | 62 | ||
| GenBank mRNA | AF543560.1 | - | 2 - 3 | 62 | ||
| BC089110.1 | - | 2 - 3 | 278 | 7311852 | 62 | |
| FQ214270.1 | - | 2 - 3 | 386 | 62 | ||
| FQ228146.1 | - | 2 - 3 | 306 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aip
Genomic Map