Assay Details
| Gene Symbol: Calm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012518.3 | NP_036650.1 | 2 - 3 | 185 | 92 | |
| GenBank mRNA | BC063187.1 | - | 280 | 6890866 | 92 | |
| FQ212716.1 | - | 251 | 92 | |||
| M16659.1 | - | 2 - 3 | 185 | 92 | ||
| X13817.1 | - | 3 - 4 | 277 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calm3
Genomic Map