Assay Details
| Gene Symbol: Gpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207592.1 | NP_997475.1 | 12 - 13 | 1128 | 85 | |
| GenBank mRNA | BC062005.1 | - | 12 - 13 | 1128 | 5599776 | 85 |
| FQ215247.1 | - | 1 - 2 | 175 | 85 | ||
| FQ216077.1 | - | 1 - 2 | 176 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpi
Genomic Map