Assay Details
| Gene Symbol: Rpl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173340.2 | NP_775462.2 | 2 - 3 | 98 | 68 | |
| GenBank mRNA | BC086382.1 | - | 2 - 3 | 98 | 7301486 | 68 |
| EU000472.1 | - | 2 - 3 | 98 | 68 | ||
| X68282.1 | - | 2 - 3 | 99 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl13a
Genomic Map