Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191621.3 | NP_001178550.1 | 5 - 6 | 1773 | 58 | |
| XM_006228576.2 | XP_006228638.1 | 5 - 6 | 1982 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k10
Genomic Map