Assay Details
| Gene Symbol: Josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106256.2 | NP_001099726.2 | 2 - 3 | 281 | 55 | |
| XM_006228996.2 | XP_006229058.1 | 3 - 4 | 563 | 55 | ||
| XM_006228997.2 | XP_006229059.1 | 3 - 4 | 560 | 55 | ||
| XM_006228998.2 | XP_006229060.1 | 2 - 3 | 502 | 55 | ||
| GenBank mRNA | CK842299.1 | - | 3 - 4 | 55 | ||
| DV214517.1 | - | 2 - 3 | 281 | 55 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Josd2
Genomic Map