Assay Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107490.3 | 6 - 7 | 937 | 90 | ||
| XM_008758933.1 | XP_008757155.1 | 7 - 8 | 4448 | 90 | ||
| XM_008758934.1 | XP_008757156.1 | 7 - 8 | 4712 | 90 | ||
| XM_008758935.1 | XP_008757157.1 | 7 - 8 | 4448 | 90 | ||
| XM_008758936.1 | XP_008757158.1 | 7 - 8 | 4448 | 90 | ||
| XM_008758937.1 | XP_008757159.1 | 7 - 8 | 4448 | 90 | ||
| XM_008758938.1 | XP_008757160.1 | 6 - 7 | 1130 | 90 | ||
| XM_008758939.1 | XP_008757161.1 | 6 - 7 | 1131 | 90 | ||
| XM_008758940.1 | XP_008757162.1 | 6 - 7 | 1131 | 90 | ||
| GenBank mRNA | CB691822.1 | - | 2 - 3 | 190 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cic
Genomic Map