Assay Details
| Gene Symbol: Trappc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109410.1 | NP_001102880.1 | 3 - 4 | 298 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc6a| Gene Symbol: Trappc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109410.1 | NP_001102880.1 | 3 - 4 | 298 | 149 | |
Set Membership: |
|