Assay Details
| Gene Symbol: Klk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107509.1 | NP_001100979.1 | 1 - 2 | 545 | 125 | |
| XM_006229049.2 | XP_006229111.1 | 2 - 3 | 146 | 125 | ||
| GenBank mRNA | AJ005641.1 | - | 1 - 2 | 213 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klk8
Genomic Map