Assay Details
| Gene Symbol: Il16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105749.1 | NP_001099219.1 | 18 - 19 | 3974 | 64 | |
| XM_006229488.2 | XP_006229550.1 | 18 - 19 | 5621 | 64 | ||
| XM_006229489.2 | XP_006229551.1 | 17 - 18 | 5518 | 64 | ||
| XM_008759599.1 | XP_008757821.1 | 18 - 19 | 7437 | 64 | ||
| XM_008759600.1 | XP_008757822.1 | 19 - 20 | 3993 | 64 | ||
| XM_008759601.1 | XP_008757823.1 | 6 - 7 | 1918 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P052835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36690 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P052835 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P052835 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P052835 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P231706 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83691 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39941 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10505 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10723367 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723358 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723353 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723356 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723370 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723365 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723369 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723354 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723357 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723368 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723355 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723352 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10723351 | - | - |
| Rat Genome 230 2.0 Array | 1376895_at | - | - |
| Rat Genome 230 2.0 Array | 1376895_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376895_at | - | - |
| Affymetrix HT Rat Focus Array | 1376895_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100047335.rn.1 | TC0100005943.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376895_PM_at | - | - |
| Rat Expression Set 230 | 1376895_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723351 | - | - |
| Rat Genome U34 Set | rc_AI137163_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21784618 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365194 | - | - |

Il16
Genomic Map